Round Developing Chamber, General Glass Blowing
Supplier: General Glass Blowing
30-1
21432-863EA 312.46 USD
21432-863
Round Developing Chamber, General Glass Blowing
TLC Development Tanks
A diameter borosilicate glass jar with flat bottom and lip
Comes with snap-on plastic cap. Holds two 10 x 20cm plates. Uses 75mL of solvent for atmosphere saturation.