Foam Biopsy Pads, BIO PLAS
Supplier: Bio Plas
6120
21150-088CS 895.04 USD
21150-088
Foam Biopsy Pads, BIO PLAS
Biopsy Pads
Pads are fabricated from cellular polyester plastic foam with air pockets throughout to allow for adequate fluid movement around the tissue