Uni-Tip™ Universal Macro Pipette Tips, Bio Plas
Supplier: Bio Plas
3250
21150-054CS 881.84 USD
21150-054
Uni-Tip™ Universal Macro Pipette Tips, Bio Plas
Pipette Tips
Designed to fit most brands of macro pipettors.
  • Made of virgin polypropylene
  • Autoclavable at 121 °C