Histo Plas™ Tissue Processing Capsules, Bio Plas
Supplier: Bio Plas
6020
21150-012CS 1076.29 USD
21150-012
Histo Plas™ Tissue Processing Capsules, Bio Plas
Embedding Cassettes Tissue Capsules
Histo Plas™ disposable tissue processing capsule is made of polypropylene
The open mesh construction affords maximum fluid exchange during processing
and the durable marking surface assures permanent identification. Simply place the tissue in the Capsule and press the cover down to lock securely.
The one piece unit is hinged, perfectly aligned and easy to use.