Unsaturation Gasoline Bottle, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
15066-10
21140-008CS 674.69 USD
21140-008
Unsaturation Gasoline Bottle, Kimble®, DWK Life Sciences
Bottles Gas Washing Bottles
This KIMAX® bottle is used in the determination of unsaturated hydrocarbons in gasoline.
Manufactured from 33 expansion, low extractable borosilicate glass.
The body of the bottle (approximately 45 ml capacity) is a 'reaction vessel' whereby an accurately measured sample is pipetted into the bottle, excess reagent is added and, after the reaction is complete, more reagent is added to raise the 'fat column' into the calibrated neck of the bottle, where results are read directly as a percentage of fat in the sample.The Neck Capacity is 10 mL (100%).
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.