PYREX® Weathering Test Tube, 100 ml, 37×200 mm, Corning
Supplier: Corning
9610-100
21116-003CS 132.42 USD
21116-003
PYREX® Weathering Test Tube, 100 ml, 37×200 mm, Corning
Tubes Test Tubes
PYREX®, borosilicate glass, conical bottom, graduated.
  • Graduated upward to 0.5 ml (0.05 ml increments)
  • 0.5 to 3 ml (0.1 ml increments)
  • 3 to 5 ml (0.5 ml increments)
  • 5 to 100 ml (1 ml increments)
It is designed for testing propane, butane and isobutane mixtures. Made with a 3 mm vent hole located 180° from the front of the tube.
Certifications: Meets TS-441-3, ASTM D-1837 and D-2158 specifications.