KIMAX® Graduated Hopkins Vaccine Centrifuge Tube, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45225-10
21060-000CS 1059.04 USD
21060-000
KIMAX® Graduated Hopkins Vaccine Centrifuge Tube, Kimble Chase
Tubes Centrifuge Tubes
KIMAX® tube is designed to standardize vaccines.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The stem is marked from 0.01 to 0.05 ml in 0.01 ml intervals, with a tolerance of ±0.0025 ml. The body has graduations at 1, 5, and 10 ml with a tolerance of ±0.20 ml. Scale and legend are durable white ceramic enamel.