KIMAX® Heavy-Duty Centrifuge Tube, Graduated, with Flathead PTFE [ST] Stopper, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45176-50
21054-190CS 622.02 USD
21054-190
KIMAX® Heavy-Duty Centrifuge Tube, Graduated, with Flathead PTFE [ST] Stopper, Kimble Chase
Tubes Centrifuge Tubes
Tube is calibrated to contain and includes a flathead, PTFE [ST]16 stopper.
  • Marking spot is blasted on both sides
  • Scale and legend are printed in permanent brown stain
Tube is graduated from 0 to 10 ml in 0.5 ml increments and from 10 to 50 ml in 1 ml increments.
Caution: Do not centrifuge with stopper in tube.