KIMAX® Heavy-Duty Graduated Centrifuge Tubes with Flat Head Stoppers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
45176-13
21054-188CS 478.68 USD
21054-188
KIMAX® Heavy-Duty Graduated Centrifuge Tubes with Flat Head Stoppers, DWK Life Sciences
Tubes Centrifuge Tubes
These tubes are manufactured from 33 expansion, low extractable borosilicate glass.
  • Calibrated to contain
  • With top finish to accept a Standard Taper stopper
  • Scale and legend are printed in permanent red stain on the 13 ml size and permanent brown stain on 50 ml size
  • Sandblasted marking spot for added durability
Certifications: Material conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Caution: Do not centrifuge with stopper in tube.