PYREX® 12 ml Heavy-Duty Conical Centrifuge Tube with Beaded Rim, Corning
Supplier: Corning
8120-12
21021-905CS 201.64 USD
21021-905
PYREX® 12 ml Heavy-Duty Conical Centrifuge Tube with Beaded Rim, Corning
Tubes Centrifuge Tubes
PYREX® borosilicate glass, conical.
These centrifuge tubes are made with increased wall thickness for greater mechanical strength.
Manufactured to the same Ext.Ø and length as standard. In order to conform to the overall Ext.Ø. and length limitations by the shields, it has been necessary to reduce the capacity of the tube.