Nunc® Centrifuge Bottle/Tube, Polypropylene, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
376813
21020-500CS 278.86 USD
21020-500
Nunc® Centrifuge Bottle/Tube, Polypropylene, Sterile, Thermo Scientific
Bottles Centrifuge Bottles
Conical, polypropylene tube fits most centrifuges with 250 or 500 ml swinging bucket rotors.
Seals tightly with a polyethylene plug-seal screw cap. The epoxy coated wire rack stores and holds six bottles for convenient handling.
Ordering information: Supplied sterile with cap. Cushion support and rack are available separately.