Corning® Centrifuge Tubes, Sterile, Polypropylene, Corning
Supplier: Corning
430776 431123
21008-771CS 399.51 USD
21008-771 89091-000
Corning® Centrifuge Tubes, Sterile, Polypropylene, Corning
Tubes Centrifuge Tubes
PP, with HDPE plug seal cap, sterile. Polyetherimide cushions.
  • Moulded graduations, conical bottom
  • Tubes are sterile and certified non pyrogenic
  • Support cushions must be used unless the rotor has appropriately shaped V-bottom holders
Ideal for applications requiring large volume centrifugation.
They are perfect for large volume centrifugation applications. They incorporate plug seal caps and have molded graduations.
Ordering information: Non sterile cushion sold separately.
Packaging: Bulk, 6/bag.