Corning® Centrifuge Tubes, 15 ml
Supplier: Corning
430055 430053 430790 430791 430766 430052 431470
21008-656CS 429.13 USD
21008-656 21008-667 21008-670 21008-671 21008-673 21008-678 89093-186
Corning® Centrifuge Tubes, 15 ml
Tubes Centrifuge Tubes
Corning® 15 ml clear centrifuge tubes are available without cap, with the advanced Corning CentriStar™ cap or the original plug seal cap.
  • Black printed graduations and a large white marking spot
  • Maximum RCF is 12000×g
  • Sterile, certified nonpyrogenic and DNase-/RNase-free
  • 95 kPA (14 psi) pressure tested – plug seal cap only
The CentriStar™ cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping. This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Ordering information: Foam racks and CentriStar™ caps (catalog no. 89093-188) can be purchased separately.
Packaging: Bulk packed 25 per resealable zip lock bag.