Drum Cradle, Wesco
Supplier: Wesco
240023 240024
21002-022EA 452.26 USD
21002-022 21002-024
Drum Cradle, Wesco
Carts
Economical, convenient way to tilt, drain and store 208.2 L (55 gal.) drums
Easy for one person to use. Drum distance to floor: 38 cm (15").
Outboard style has four rigid polyolefin wheels. Inboard style features two swivel, two rigid polyolefin wheels.
Packaging: Shipped unassembled for quick, easy assembly.