Upchurch Scientific® Fittings Kits, IDEX Health and Science
Supplier: Upchurch Scientific
1356 1453 1456 1454 1404 1451 1457
53498-532EA 589.1 USD
53498-532 53500-610 21001-110 21001-116 21511-370 21001-112 53498-566 53500-666 21511-368 21511-372 53500-848 21001-114
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Fittings and Connectors
MicroTight™ Tubing Sleeves facilitate capillary tubing connections with our MicroTight accessories
Manufacturer Specific Fittings Kits are customized for specific manufacturer’s legacy HPLC systems.
IDEX Health & Science manufactures a large selection of kits, each containing an assortment of products designed to make your life easier and to save you money. Our kits range from our Super Kit – which contains a collection of many products you’ll use to accessorize your systems – to more specialized kits like our FIA kit, offering products designed to help you with flow injection analyses.
Sleeves feature an outer diameter of .025" and are available in variety of inner diameters for a broad-range compatibility with a wide assortment of capillary tubing sizes. Manufactured from PEEK polymer, these sleeves carry an upper temperature threshold of 125 °C. For proper sleeve use, choose a sleeve with an inner diameter .001"-.002" (25-50 µm) larger than the outer diameter of your capillary tubing. Then slip the sleeve over the flow path tubing, such that the tubing extends all the way through the sleeve, but not beyond the end of the sleeve. Choose the correct fitting that corresponds with your receiving port, slide it over the sleeved flow path tubing and connect as normal.
Certifications: ROHS Certified