SP Bel-Art Microcentrifuge Tube, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19929-0000
20901-925PK 62.11 USD
20901-925
SP Bel-Art Microcentrifuge Tube, Bel-Art Products, a part of SP
Tubes Microcentrifuge Tubes
These micro sample tubes have a captive plug and will withstand the forces of a microcentrifuge.
  • Captive plug design
  • Polypropylene construction
  • Autoclavable
Polypropylene tubes are steam autoclavable at 121 °C (250 °F).