Distilling Adapter, Iowa State, HGF GLASS INC
Supplier: HGF Glass
210200-1
20203-004EA 255.08 USD
20203-004
Distilling Adapter, Iowa State, HGF GLASS INC
Distillation Components Distillation Adapters
Distilling adapter is a component of Iowa state distillation unit which have been used for determinations in the citrus industry but also applicable for other distilling procedures.