Micro Tube Tool, Bio Plas
Supplier: Bio Plas
4000
20170-688CS 438.7 USD
20170-688
Micro Tube Tool, Bio Plas
Tubes Microtubes
The Micro Tube Tool prevents fatigue and saves wear on fingers and thumbs.
  • Opens tight-fitting Snap Caps
  • Allows removal of Screw Caps while tubes are still in the rack
  • Opens Screw-Cap tubes and any size Snap Cap tubes
The handy polypropylene Micro Tube Tool opens Snap-Cap and Screw Cap microcentrifuge tubes easily, prevents fatigue, and saves wear on user's fingers and thumbs.
This innovative tool makes it a breeze to open tight-fitting Snap Caps, and allows users to remove Screw Caps while tubes are still in the rack. Convenient and economical, the Micro Tube Tool puts an end to difficult maneuvering and straining to open Screw-Cap tubes and any size Snap Cap tubes.
Made in the USA.