Nb.BtsI
Supplier: New England Biolabs (NEB)
R0707S
200067-224EA 82 USD
200067-224
Nb.BtsI
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that expresses only the large subunit of the BtsI restriction gene from Bacillus thermoglucosidasius (X.Pan).