Magnetic Separation Rack, 12-Tube, New England Biolabs
Supplier: New England Biolabs (NEB)
S1509S
200064-492EA 399 USD
200064-492
Magnetic Separation Rack, 12-Tube, New England Biolabs
Racks Magnetic Separation Racks
The Magnetic Separation Racks are designed to be used for small-scale separations using magnetic particles
The magnets are located on the sides of the racks, resulting in minimal sample loss during pipetting.