Magnetic Separation Rack, 50 ml, New England Biolabs
Supplier: New England Biolabs (NEB)
S1507S
200064-490EA 358 USD
200064-490
Magnetic Separation Rack, 50 ml, New England Biolabs
Racks Magnetic Separation Racks
The Magnetic Separation Racks are designed to be used for small-scale separations using magnetic particles
The magnets are located on the sides of the racks, resulting in minimal sample loss during pipetting.