Anti-SYN2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
VAS-SV061-E
200063-318EA 367.33 USD
200063-318
Anti-SYN2 Rabbit Polyclonal Antibody
Antibodies
Synapsin I, II and III belong to a family of neuron-specific phosphoproteins which maintain a reserve pool of vesicles in the vicinity of the active zone by tethering synaptic vesicles to each other and to an Actin-based cytoskeleton meshwork. They have been shown to regulate neurotransmitter release in mature synapses and to accelerate development of the nervous system.

  • For Immunoprecipitation, Western Blot

Recognizes mouse and rat synapsin II. Detects a band of ~74kDa (synapsin Iia) and ~54kDa (synapsin Iib) by Western blot.

Type: Primary
Antigen: SYN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR