Anti-CANX Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-865-D ADI-SPA-865-F
200063-276EA 236.53 USD
200063-276 200062-780
Anti-CANX Rabbit Polyclonal Antibody
Antibodies
Calnexin (CNX), an unglycosylated resident ER transmembrane protein, together with Calreticulin (CRT), plays a key role in glycoprotein folding and its control within the ER, by interacting with folding intermediates via their monoglucosylated glycans. Calnexin associates with newly synthesized monomeric glycoproteins and only recognizes glycoproteins when they are incompletely folded. Furthermore, Calnexin has been demonstrated to function as a molecular chaperone capable of interacting with polypeptide segments of folding glycoproteins.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, C. elegans, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Calnexin. Detects a band of ~90kDa by Western blot.

Type: Primary
Antigen: CANX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR