D-Gluconic acid 50% (w/w) in water

Supplier: Thermo Scientific Chemicals

Synonyms: Gluconic acid, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid

AC11989-0025
200057-670EA 41.4 USD
200057-670 200057-672 103516-414
D-Gluconic acid 50% (w/w) in water
D-Gluconic acid
Formula: C₆H₁₂O₇
MW: 196.16 g/mol
Melting Pt: < 15 °C
Density: 1.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004240
CAS Number: 526-95-4
EINECS: 208-401-4
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear yellow to brown-yellow Solution
Titration excess NaOH 49 to 55 %
Reducing sugars ≤3 %
Titration Argentometric ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR