Rhodium 5% on alumina, powder

Supplier: Thermo Scientific Chemicals

AC19957-0250
200026-684EA 443 USD
200026-684 200056-250
Rhodium 5% on alumina, powder
Rhodium
Formula: Rh
Boiling Pt: 3147 °C (760 mmHg)
Melting Pt: 2072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Brown or grey to black Powder
Assay 5 % (Rh) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR