1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Mesitylsulphonyl)-3-nitro-1H-1,2,4-triazole, 1-(Mesitylsulfonyl)-3-nitro-1H-1,2,4-triazole

AC22131-0050
200015-440EA 106 USD
200015-440 200045-572
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole 98%
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole

Appearance: White to yellow Powder or crystals

Formula: C₁₁H₁₂N₄O₄S
MW: 296.301 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Freezer
MDL Number: MFCD00009754
CAS Number: 74257-00-4

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 131 to 139 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR