2-Methylindoline 99+%

Supplier: Thermo Scientific Chemicals
Warning

AC27588-1000
200041-180EA 105.86 USD
200041-180 200028-878
2-Methylindoline 99+%
2-Methylindoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 228
Melting Pt: –51 °C
Density: 1.02 g/cm³
Flash Pt: 103 °C
MDL Number: MFCD00005706
CAS Number: 6872-06-6
EINECS: 229-971-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR