2-Methyl-2-oxazoline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methyl-4,5-dihydrooxazole

AC15997-1000
200046-974EA 282 USD
200046-974 200034-892
2-Methyl-2-oxazoline 99%
2-Methyl-2-oxazoline
Formula: C₄H₇NO
MW: 85.11 g/mol
Density: 1 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00005298
CAS Number: 1120-64-5
EINECS: 214-316-3
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR