2,5-Dimercapto-1,3,4-thiadiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,4-Thiadiazole-2,5-dithiol

AC11531-1000
200022-246EA 30.3 USD
200022-246 200046-466 200034-420
2,5-Dimercapto-1,3,4-thiadiazole 98%
2,5-Dimercapto-1,3,4-thiadiazole
Formula: C₂H₂N₂S₃
MW: 150.25 g/mol
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00003103
CAS Number: 1072-71-5
EINECS: 214-014-1

Order Now

Specification Test Results

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Melting point 156 to 162 °C (Dec)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR