2-Hydrazinobenzoic acid hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

AC41180-0250
200026-290EA 81.37 USD
200026-290 200022-688
2-Hydrazinobenzoic acid hydrochloride 99%
2-Hydrazinobenzoic acid hydrochloride
Formula: C₇H₉ClN₂O₂
MW: 188.61 g/mol
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012931
CAS Number: 52356-01-1
EINECS: 257-869-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR