Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 90%

Supplier: Thermo Scientific Chemicals

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

AC22631-0010
200021-136EA 272.91 USD
200021-136
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 90%
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
Melting Pt: >140 °C (dec.)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00044310
CAS Number: 19817-92-6
EINECS: 243-347-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤15 % (105 °C, 5 hrs)
HPLC ≥88 %
UV Lambda max 262 nm (+/- 2 nm)
pH 3.0 to 7.0 (0.5 % in water)
Absorbance scale 0.70 to 0.80 (A250/A260)
Chloride (Cl) ≤2000 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR