Industrial Gloves, MaxiFlex® Ultimate™ 34-874, Protective Industrial Products
Supplier: PIP
34-874/L 34-874/XL 34-874/M 34-874/S
20002-352CS 2044.69 USD
20002-352 20002-354 20002-356 20002-358
Industrial Gloves, MaxiFlex® Ultimate™ 34-874, Protective Industrial Products
Gloves
The MaxiFlex® Ultimate™ has become the benchmark for precision handling in dry environments.
  • Soft liner made from Nylon and Spandex
  • Micro-foam NBR (Nitrile Butadiene Rubber) palm coating for controlled grip
  • Silicone free
  • FDA compliant, Food contact approved (EU)
  • Touchscreen compatible
The patented micro foam nitrile coating offers 360° breathability, reduces hand fatigue and increases comfort. Gloves are washable.
MaxiFlex® has outstanding durability withstanding 18.000 abrasive cycles per millimetre thanks to the use of the DURAtech® coating technology in the coating construction.
Typical applications include: General assembly, micro-engineering, automotive components, painting, and horticulture.