Homophthalic anhydride 98%

Supplier: Thermo Scientific Chemicals
Warning

AC15630-0250
200019-330EA 34.5 USD
200019-330 200019-328 103510-660
Homophthalic anhydride 98%
Homophthalic anhydride
Formula: C₉H₆O₃
MW: 162.14 g/mol
Melting Pt: 140…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR