1-Bromo-3-methylbutane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylbutylbromide

AC40305-5000
200027-818EA 18.8 USD
200027-818 200015-712 200008-596
1-Bromo-3-methylbutane 99%
1-Bromo-3-methylbutane
Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 119
Melting Pt: -112
Density: 1.2 g/cm³
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000253
CAS Number: 107-82-4
EINECS: 203-522-9
UN: 2341
ADR: 3 III
Merck Index: 13,05130

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4399 to 1.4419 (20 °C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥98.5 %
Acidity ≤0.1 % (as HBr)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR