4-Morpholinoaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4-Morpholinyl)aniline

AC18865-0050
200014-080EA 67.09 USD
200014-080 102776-740
4-Morpholinoaniline 98+%
4-Morpholinoaniline
Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
Melting Pt: 130…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00006169
CAS Number: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR