Indole-3-carbaldehyd 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde

AC15664-0250
200025-572EA 270 USD
200025-572 200012-742 103510-742
Indole-3-carbaldehyd 97%
Indole-3-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 193…199 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00005622
CAS Number: 487-89-8
EINECS: 207-665-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR