Premium Grade Cotton Canvas Single Palm Gloves with Plasticized Gauntlet Cuff, PIP®, Protective Industrial Products
Supplier: PIP
90-910GA
20001-974CS 2216.52 USD
20001-974
Premium Grade Cotton Canvas Single Palm Gloves with Plasticized Gauntlet Cuff, PIP®, Protective Industrial Products
Gloves
Multi-purpose fabric gloves used for light protection in many general applications.
  • Natural cotton canvas keeps hands cool and protected
  • Plasticized gauntlet cuff for added forearm protection and allows for easy on and off
  • Clute cut provides a roomy fit
  • Straight thumb increases comfort in closed-fisted work
Heavy weight provides enhanced protection and extended wear.