Zinc 99+%, mossy

Supplier: Thermo Scientific Chemicals

AC20145-5000
200008-092EA 61.38 USD
200008-092 103509-632 103509-634
Zinc 99+%, mossy
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Appearance Silver grey Mossy flakes
Total trace metal impurities ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR