Cytidine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

AC11181-0100
200006-176EA 74.52 USD
200006-176 200030-832 103508-746
Cytidine 99+%
Cytidine
Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Melting Pt: 210…217 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006545
CAS Number: 65-46-3
EINECS: 200-610-9

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 210 to 217 °C (Dec)
Loss on drying ≤0.5 % (105 °C, 5 hrs)
Sulfated ash ≤0.1 % (800 °C)
HPLC ≥99.0 %
UV spectrophotometry molar absorptivity 13000 +/- 2%, on dry basis
UV transmittance ≥98 % (c=1, H2O, 430 nm) 1 cm cell
Ratio of Absorbances 2.05 to 2.15 (A280/A260)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR