2-Chloro-2-methylpropane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl chloride

AC10950-0250
200000-216EA 49.8 USD
200000-216 200009-278 200005-010
2-Chloro-2-methylpropane 99%
2-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 51
Melting Pt: -28
Density: 0.85 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3 II
Merck Index: 13,01561

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3838 to 1.3858 (20 °C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR