Acetic anhydride 97+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetyl oxide, Acetyl acetate

AC42323-5000
200004-064EA 77.42 USD
200004-064
Acetic anhydride 97+% ACS
Acetic anhydride
Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 139.9 °C (1013 hPa)
Melting Pt: –73.1 °C
Density: 1.087 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8,II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
GC ≥97.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.003 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤2 ppm
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR