Acetic anhydride, anhydrous 99+%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetyl oxide, Acetyl acetate

AC14949-0010
200042-400EA 547 USD
200042-400 200001-124
Acetic anhydride, anhydrous 99+%, pure
Acetic anhydride
Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 139.9 °C (1013 hPa)
Melting Pt: –73.1 °C
Density: 1.087 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8,II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR