L(-)-Tryptophan 99%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

AC14059-0250
200006-766EA 107 USD
200006-766 200031-662 200006-764 200000-982 103510-442
L(-)-Tryptophan 99%
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 290 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now

Specification Test Results

Appearance White to yellowish-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105 °C)
Specific optical rotation -29.0° to -33.0° (20 °C, 589 nm) (C=1, H2O)
Titration with HClO4 98.5 to 101.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR