G-Tek® GP™ Seamless Knit Nylon Gloves with PU Coating, Protective Industrial Products
Supplier: PIP
33-G125/S
20000-713CS 1315.33 USD
20000-713 20000-715
G-Tek® GP™ Seamless Knit Nylon Gloves with PU Coating, Protective Industrial Products
Gloves
Combination of comfortable nylon shell, and a protective PU coating makes this glove ideal for use in electronic and computer assembly, quality control, inspection and general assembly.
  • Excellent abrasion resistance
  • Knit wrist to prevent dirt and debris from entering the glove
  • Breathable back for comfort
  • Pair packed for ease of distribution
  • Continuous knitted cuffs
The seamless knit nylon shell offers increased comfort, finger dexterity and breathability. The polyurethane coated palm and fingertips provide a secure grip in dry and slightly wet/oily conditions, while also offering high dexterity and tactile sensitivity.
Certifications: EN 388: 4131. ANSI Cut Level 1. CFIA Approved.