Steam Sterilization Indicator Labels, NAMSA®
Supplier: NAMSA
NMCPI-S01
19872-046EA 259.4 USD
19872-046
Steam Sterilization Indicator Labels, NAMSA®
Sterilization Indicators
Self-adhesive, nontoxic, chemical process indicators change from blue to pink upon exposure to 10 minutes of saturated steam at 121 °C (250 °F) or 2 minutes at 134 °C (273 °F).
Longer exposure will result in a brighter shade of pink. The blue to pink color transition is sensitive to time, temperature, and the presence of saturated steam. Water-based ink is made without the use of heavy metals. Indicators are stable for at least 18 months from date of manufacture when they are stored at room temperature.
Certifications: Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.