Quick Set™ 404™ Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
135465
19807-542CS 441.35 USD
19807-542
Quick Set™ 404™ Instant Adhesive, Loctite®, Henkel
Adhesives
Quick Set™ 404™ adhesive is a general-purpose, medium-viscosity, instant adhesive.
  • 3,500 psi tensile strength
  • Fills gaps up to 0.012 cm (0.005")
Adhesive is ideal for bonding rubber parts, such as belts, bumpers, cushions, and O-rings.