Permabond® QFS16 Ozone-Safe Accelerator
Supplier: Permabond
QFS16-4OUNCE
19806-014CS 258.18 USD
19806-014
Permabond® QFS16 Ozone-Safe Accelerator
Adhesives
This accelerator is for use with all Permabond® cyanoacrylate adhesives.
  • Increases gap-filling properties and cure time
  • Pump spray bottle
Accelerator is used to increase gap-filling properties and cure time.