Super Erecta Shelf® Stem Caster Carts, Metro™
Supplier: Metro
N436BC N356EC N466EC N536EC N366EC N536BBR N456BC N466BC N336EC N456EC N556EC N436EC N566EC N336EBR N336BC N336BC N356BC N356BC N536BC N536BC N556BC N566BC N566BC N366BC N366BC
19766-028EA 963.41 USD
19766-028 19766-624 19766-632 19766-634 19766-626 82002-472 19766-030 19766-312 19766-622 19766-630 19766-636 19766-628 19766-638 82002-470 19766-018 19766-020 19766-022 19766-024 19766-032 19766-034 19766-038 19766-040 19766-042 19766-310 19766-026
Super Erecta Shelf® Stem Caster Carts, Metro™
Carts
These four-shelf carts with casters are designed for safe transport and storage
Chrome carts feature chrome-plated wire shelving and posts. Super Erecta Brite* carts feature zinc-plated shelves and chrome-plated posts. Both finishes are ideal for use in dry environments. Open wire shelf design minimizes dust and increases air circulation and visibility. Shelves are adjustable at 2.5cm (1") increments and utilize snap-on tapered “sleeves” to secure them to posts.
Ordering information: Carts are shipped knocked down for easy assembly. Each cart includes four shelves, posts, plastic split sleeves, donut bumpers, and designated casters. For other cart sizes and accessories, contact your Avantor sales representative.