Super Erecta® Wire Shelving, Stainless Steel, Metro™
Supplier: Metro
3072NS 3048NS 3636NS 3660NS 2130NS 2154NS 2160NS 2172NS 3060NS 3036NS 3648NS 3672NS 1424NS 1430NS 1448NS 1460NS 1472NS 1824NS 1848NS 1860NS 2148NS 2460NS 1854NS 1436NS 1442NS 1830NS 1836NS 1842NS 1872NS 2124NS 2136NS 2142NS 2424NS 2430NS 2436NS 2442NS 2448NS 2454NS 2472NS
19767-054EA 1226.04 USD
19767-054 19767-052 19767-056 19767-060 19766-520 19766-526 19766-528 19766-530 19766-810 19767-050 19767-058 19767-062 19766-072 19766-074 19766-084 19766-088 19766-092 19766-098 19766-122 19766-140 19766-168 19766-204 19766-386 19766-504 19766-506 19766-508 19766-510 19766-512 19766-516 19766-518 19766-522 19766-524 19766-532 19766-534 19766-536 19766-538 19766-540 19766-542 19766-544
Super Erecta® Wire Shelving, Stainless Steel, Metro™
Furniture Shelves
Type 304 stainless steel shelves provide maximum protection in corrosive environments.
  • Open-wire design
  • Easy shelf adjustment
  • Double grooves to aid shelf alignment
Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items
Shelves up to 121.9 cm (48) in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") can support up to 272kg (600lbs.) per level.
Note: The 35.6 cm (14) wide shelves require the use of footplates (see 19766-814 series) when used as a freestanding unit. The actual length of the shelves is 3.2 mm (1/8") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap.
Easy shelf adjustment is possible through the use of SiteSelect® stationary or mobile posts (sold separately), which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1) increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each individual shelf and post is sold separately. To order posts or accessories, contact your Avantor representative for more information. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts.