Mayo-Style Instrument Tray, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
80130
19760-060CS 596.71 USD
19760-060
Mayo-Style Instrument Tray, Medegen Medical Products
Trays/Dishes/Pots Instrument Trays/Dishes
This instrument tray has a satin finish, smooth rounded corners, and open beads for easy cleaning
Can be steam sterilized to 135°C (275°F).
Certifications: NSA (National Sanitation Association) approved.